Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF2-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028043.2 | 2 | 704 | |||
NR_133657.1 | 2 | 593 | ||||
GenBank mRNA | AL521915.3 | - | 2 | 697 | ||
AL530868.3 | - | 2 | 598 | |||
AL545164.3 | - | 2 | 559 |