Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016436.4 | NP_057520.2 | 10 | 1454 | ||
XM_005260421.2 | XP_005260478.1 | 7 | 1064 | |||
XM_011528843.1 | XP_011527145.1 | 9 | 1055 | |||
XM_011528844.1 | XP_011527146.1 | 7 | 1126 | |||
XM_017027864.1 | XP_016883353.1 | 11 | 1536 | |||
XM_017027865.1 | XP_016883354.1 | 11 | 1737 | |||
XM_017027866.1 | XP_016883355.1 | 10 | 1657 | |||
XM_017027867.1 | XP_016883356.1 | 10 | 1492 | |||
XM_017027868.1 | XP_016883357.1 | 8 | 1278 | |||
XM_017027869.1 | XP_016883358.1 | 10 | 1451 | |||
XM_017027870.1 | XP_016883359.1 | 10 | 1457 | |||
XM_017027871.1 | XP_016883360.1 | 11 | 1537 | |||
GenBank mRNA | AF220416.1 | - | 11 | 1357 | ||
AF258787.1 | - | 10 | 1440 | |||
AF348207.1 | - | 9 | 1379 | |||
AK054597.1 | - | 11 | 1711 | |||
AK314503.1 | - | 10 | 1418 | |||
AY027523.1 | - | 9 | 1379 | |||
AY134747.1 | - | 10 | 1431 | |||
BC006415.1 | - | 9 | 1314 | |||
BC016948.1 | - | 10 | 1407 | |||
BC048210.1 | - | 11 | 1487 | |||
BC080598.1 | - | 10 | 1418 | |||
BC093405.1 | - | 10 | 1454 | |||
BC150178.1 | - | 10 | 1420 | 40148074 |