Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016436.4 | NP_057520.2 | 16 | 2524 | ||
XM_005260421.2 | XP_005260478.1 | 13 | 2134 | |||
XM_011528843.1 | XP_011527145.1 | 15 | 2125 | |||
XM_011528844.1 | XP_011527146.1 | 13 | 2196 | |||
XM_017027864.1 | XP_016883353.1 | 17 | 2606 | |||
XM_017027865.1 | XP_016883354.1 | 17 | 2807 | |||
XM_017027866.1 | XP_016883355.1 | 16 | 2727 | |||
XM_017027867.1 | XP_016883356.1 | 16 | 2562 | |||
XM_017027868.1 | XP_016883357.1 | 14 | 2348 | |||
XM_017027869.1 | XP_016883358.1 | 16 | 2521 | |||
XM_017027870.1 | XP_016883359.1 | 16 | 2527 | |||
XM_017027871.1 | XP_016883360.1 | 17 | 2607 | |||
GenBank mRNA | AK097498.1 | - | 5 | 727 | ||
AK314503.1 | - | 16 | 2488 | |||
AY027523.1 | - | 15 | 2449 | |||
BC150178.1 | - | 16 | 2490 | 40148074 |