Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MZB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016459.3 | NP_057543.2 | 4 | 645 | ||
GenBank mRNA | AA837360.1 | - | 1 | 261 | ||
AF151024.1 | - | 4 | 724 | |||
AF338109.1 | - | 4 | 826 | |||
AK292706.1 | - | 4 | 620 | |||
BC009931.2 | - | 3 | 499 | 2989349 | ||
BC021275.2 | - | 4 | 626 | 4853250 |