Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHISA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272065.1 | NP_001258994.1 | 5 | 615 | ||
NM_001272066.1 | NP_001258995.1 | 5 | 602 | |||
NM_001272067.1 | NP_001258996.1 | 5 | 797 | |||
NM_001272068.1 | NP_001258997.1 | 6 | 834 | |||
NM_001272082.1 | NP_001259011.1 | 3 | 491 | |||
NM_001272083.1 | NP_001259012.1 | 3 | 491 | |||
NM_016479.4 | NP_057563.3 | 5 | 636 | |||
XM_005265205.1 | XP_005265262.1 | 6 | 884 | |||
XM_006713188.2 | XP_006713251.1 | 5 | 537 | |||
XM_011533796.1 | XP_011532098.1 | 6 | 626 | |||
GenBank mRNA | AB097007.1 | - | 3 | 255 | ||
AF151051.1 | - | 3 | 302 | |||
AF520698.1 | - | 5 | 615 | |||
AK027728.1 | - | 1017 | ||||
AK056328.1 | - | 5 | 778 | |||
AK075441.1 | - | 5 | 551 | |||
AK124652.1 | - | 6 | 834 | |||
AK298799.1 | - | 3 | 316 | |||
AK298842.1 | - | 3 | 266 | |||
AK301191.1 | - | 3 | 550 | |||
AK310083.1 | - | 2 | 439 | |||
AL117413.1 | - | 3 | 262 | |||
AL832568.1 | - | 3 | 298 | |||
BC001463.2 | - | 3 | 306 | 3138644 | ||
BC020279.2 | - | 3 | 256 | |||
BI552637.1 | - | 5 | 640 | |||
BX336842.2 | - | 5 | 519 | |||
BX344125.2 | - | 5 | 562 | |||
BX379507.2 | - | 5 | 508 | |||
DA046900.1 | - | 3 | 491 |