Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142468.2 | NP_001135940.1 | 3 | 1125 | ||
NM_001142470.2 | NP_001135942.1 | 4 | 1208 | |||
NM_016487.4 | NP_057571.1 | 3 | 599 | |||
XM_011535876.2 | XP_011534178.1 | 4 | 703 | |||
XM_011535877.2 | XP_011534179.1 | 4 | 651 | |||
XM_017010919.1 | XP_016866408.1 | 3 | 598 | |||
XM_017010920.1 | XP_016866409.1 | 3 | 1091 | |||
GenBank mRNA | AF151064.1 | - | 3 | 555 | ||
AI193526.1 | - | 2 | 345 | |||
AK091564.1 | - | 3 | 1084 | |||
BC010899.2 | - | 2 | 501 | 4078296 | ||
BX333652.2 | - | 4 | 811 |