Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173990.2 | NP_001167461.1 | 4 | 593 | ||
NM_001173991.2 | NP_001167462.1 | 4 | 593 | |||
NM_016499.5 | NP_057583.2 | 4 | 574 | |||
XM_005274039.3 | XP_005274096.1 | 5 | 1414 | |||
XM_017017882.1 | XP_016873371.1 | 4 | 1280 | |||
GenBank mRNA | AF151078.1 | - | 4 | 334 | ||
AK303687.1 | - | 4 | 593 | |||
BC011010.1 | - | 4 | 364 | 4286625 | ||
BG759319.1 | - | 4 | 573 | |||
CR457166.1 | - | 2 | 138 | |||
DN998400.1 | - | 4 | 491 | |||
KU159424.1 | - | 4 | 380 |