Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300888.1 | NP_001287817.1 | 3 | 347 | ||
NM_016500.3 | NP_057584.2 | 3 | 340 | |||
GenBank mRNA | BC001220.1 | - | 3 | 340 | 3528586 | |
BC051894.1 | - | 3 | 202 | 6528202 | ||
BM817702.1 | - | 3 | 236 | |||
CD704296.1 | - | 3 | 241 |