Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297748.1 | NP_001284677.1 | 2 | 414 | ||
NM_001297750.1 | NP_001284679.1 | 4 | 611 | |||
NM_001297751.1 | NP_001284680.1 | 3 | 555 | |||
NM_016511.3 | NP_057595.2 | 3 | 513 | |||
XM_011520687.2 | XP_011518989.1 | 3 | 513 | |||
GenBank mRNA | AF200949.1 | - | 3 | 451 | ||
AK075114.1 | - | 3 | 433 | |||
AK091989.1 | - | 2 | 365 | |||
AK291584.1 | - | 3 | 401 | |||
AK313589.1 | - | 3 | 431 | |||
AY358587.1 | - | 3 | 513 | |||
BC039072.1 | - | 3 | 419 | 5178017 | ||
BC067746.1 | - | 3 | 433 | 30337675 | ||
BM920364.1 | - | 453 | ||||
BQ722745.1 | - | 3 | 442 | |||
BX339075.2 | - | 3 | 451 | |||
BX380212.2 | - | 4 | 468 |