Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIPOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016518.2 | NP_057602.2 | 2, 3 | 587 | ||
GenBank mRNA | AF134593.1 | - | 2, 3 | 304 | ||
AF136970.1 | - | 2, 3 | 273 | |||
AK022773.1 | - | 2, 3 | 352 | |||
AK027498.1 | - | 2, 3 | 311 | |||
BC008960.2 | - | 2, 3 | 587 | 2961289 | ||
BC027622.2 | - | 2, 3 | 323 | 4797450 | ||
CR457155.1 | - | 2, 3 | 261 |