Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIPOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016518.2 | NP_057602.2 | 6 | 1273 | ||
XM_017024722.1 | XP_016880211.1 | 5 | 872 | |||
GenBank mRNA | AF134593.1 | - | 6 | 990 | ||
AF136970.1 | - | 6 | 959 | |||
AK022773.1 | - | 6 | 1038 | |||
AK027498.1 | - | 6 | 997 | |||
BC008960.2 | - | 6 | 1273 | 2961289 | ||
BC027622.2 | - | 6 | 1009 | 4797450 | ||
CR457155.1 | - | 6 | 947 |