Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171201.1 | NP_001164672.1 | 3 | 690 | ||
NM_001171202.1 | NP_001164673.1 | 3 | 606 | |||
NM_001171203.2 | NP_001164674.1 | 5 | 886 | |||
NM_001171204.2 | NP_001164675.1 | 5 | 854 | |||
NM_016525.4 | NP_057609.2 | 4 | 731 | |||
NR_033243.2 | 5 | 847 | ||||
XM_006716779.3 | XP_006716842.1 | 4 | 757 | |||
XM_011517898.2 | XP_011516200.1 | 3 | 524 | |||
XM_011517899.1 | XP_011516201.1 | 5 | 788 | |||
XM_011517900.1 | XP_011516202.1 | 4 | 663 | |||
XM_017014801.1 | XP_016870290.1 | 5 | 1123 | |||
XM_017014802.1 | XP_016870291.1 | 3 | 480 | |||
GenBank mRNA | AF222043.2 | - | 4 | 660 | ||
AK074426.1 | - | 4 | 724 | |||
AK074724.1 | - | 2 | 469 | |||
AK074745.1 | - | 4 | 691 | |||
AK074812.1 | - | 4 | 731 | |||
AK074969.1 | - | 5 | 833 | |||
AK074995.1 | - | 4 | 695 | |||
AK223235.1 | - | 4 | 686 | |||
AK295482.1 | - | 3 | 566 | |||
AK303711.1 | - | 3 | 690 | |||
AK309329.1 | - | 3 | 561 | |||
AL136733.1 | - | 4 | 524 | |||
BC020950.1 | - | 4 | 693 | 3851813 | ||
BC098141.1 | - | 4 | 627 | 40011247 | ||
BC098316.1 | - | 4 | 627 | 40011248 | ||
BC099726.1 | - | 4 | 627 | 40011249 | ||
BC100668.1 | - | 4 | 627 | 40011243 | ||
CR533551.1 | - | 3 | 488 |