Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016562.3 | NP_057646.1 | 3 | 166 | ||
GenBank mRNA | AB445659.1 | - | 1 | 27 | ||
AF240467.1 | - | 3 | 162 | |||
AF245702.1 | - | 3 | 160 | |||
AK313858.1 | - | 3 | 177 | |||
AY358292.1 | - | 2 | 111 | |||
BC033651.1 | - | 3 | 177 | 5582912 |