Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243193.1 | NP_001230122.1 | 5 | 548 | ||
NM_016583.3 | NP_057667.1 | 5 | 548 | |||
NM_130852.2 | NP_570913.1 | 5 | 548 | |||
GenBank mRNA | AB024937.1 | - | 5 | 545 | ||
AF158745.1 | - | 5 | 539 | |||
AF172993.1 | - | 5 | 525 | |||
AF417256.1 | - | 5 | 548 | |||
AF417257.1 | - | 5 | 548 | |||
AF439448.1 | - | 5 | 532 | |||
AK292778.1 | - | 5 | 547 | |||
AY359020.1 | - | 5 | 549 | |||
BC012549.1 | - | 5 | 552 | 4246419 | ||
DQ846869.1 | - | 3 | 436 |