Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101800.2 | NP_001095270.1 | 4 | 805 | ||
NM_001101801.2 | NP_001095271.1 | 3 | 613 | |||
NM_016603.3 | NP_057687.2 | 4 | 805 | |||
XM_005272007.1 | XP_005272064.1 | 4 | 805 | |||
XM_006714647.1 | XP_006714710.1 | 4 | 805 | |||
XM_006714648.1 | XP_006714711.1 | 4 | 557 | |||
XM_006714649.1 | XP_006714712.1 | 3 | 613 | |||
XM_011543448.1 | XP_011541750.1 | 5 | 598 | |||
XM_011543449.1 | XP_011541751.1 | 5 | 902 | |||
XM_011543450.1 | XP_011541752.1 | 4 | 514 | |||
XM_017009549.1 | XP_016865038.1 | 4 | 830 | |||
XM_017009550.1 | XP_016865039.1 | 4 | 514 | |||
XM_017009551.1 | XP_016865040.1 | 3 | 613 | |||
GenBank mRNA | AF251038.1 | - | 4 | 724 | ||
AK314224.1 | - | 4 | 781 | |||
BC041335.1 | - | 3 | 591 | |||
BC048965.1 | - | 4 | 777 | 5265611 | ||
BC056887.1 | - | 3 | 380 | 6148274 | ||
BU183000.1 | - | 3 | 379 |