Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304526.1 | NP_001291455.1 | 4 | 557 | ||
NM_016618.2 | NP_057702.1 | 4 | 618 | |||
XM_017004292.1 | XP_016859781.1 | 4 | 919 | |||
GenBank mRNA | AF208845.1 | - | 4 | 546 | ||
AF217520.1 | - | 4 | 557 | |||
AK025986.1 | - | 296 | ||||
BC015927.1 | - | 4 | 509 | 3908532 | ||
BC107580.1 | - | 4 | 584 | 4668988 | ||
EF413023.1 | - | 1 | 152 |