Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016628.4 | NP_057712.2 | 5 | 492 | ||
NM_100264.2 | NP_567822.1 | 5 | 783 | |||
NM_100486.3 | NP_567823.1 | 5 | 492 | |||
XM_011519491.2 | XP_011517793.1 | 5 | 1093 | |||
XM_017016315.1 | XP_016871804.1 | 5 | 603 | |||
XM_017016316.1 | XP_016871805.1 | 5 | 1093 | |||
XM_017016317.1 | XP_016871806.1 | 5 | 603 | |||
XM_017016318.1 | XP_016871807.1 | 5 | 603 | |||
GenBank mRNA | AB058747.1 | - | 3 | 202 | ||
AF116666.2 | - | 6 | 557 | |||
AK055852.1 | - | 5 | 600 | |||
AK124104.1 | - | 1 | 1022 | |||
AK223600.1 | - | 5 | 490 | |||
AK225846.1 | - | 5 | 596 | |||
AK290174.1 | - | 5 | 497 | |||
AK297460.1 | - | 5 | 405 | |||
AL713727.1 | - | 3 | 622 | |||
BC004258.1 | - | 5 | 483 | 3347345 | ||
BI226998.1 | - | 5 | 380 |