Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPG21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127889.4 | NP_001121361.1 | 8 | 816 | ||
NM_001127890.4 | NP_001121362.1 | 7 | 916 | |||
NM_016630.6 | NP_057714.1 | 8 | 997 | |||
XM_005254437.4 | XP_005254494.1 | 9 | 1030 | |||
XM_017022297.1 | XP_016877786.1 | 8 | 1365 | |||
XM_017022298.1 | XP_016877787.1 | 9 | 1455 | |||
GenBank mRNA | AF208861.1 | - | 7 | 712 | ||
AF212231.1 | - | 8 | 784 | |||
AK123725.1 | - | 6 | 1229 | |||
AK172849.1 | - | 8 | 829 | |||
AK225527.1 | - | 8 | 829 | |||
AK301362.1 | - | 7 | 881 | |||
AK312943.1 | - | 8 | 832 | |||
BC000244.1 | - | 8 | 867 | 3357388 | ||
BI258560.1 | - | 2 | 59 |