Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317990.1 | NP_001304919.1 | 2 | 548 | ||
NM_015898.3 | NP_056982.1 | 2 | 712 | |||
XM_005259570.4 | XP_005259627.3 | 2 | 1634 | |||
XM_005259571.4 | XP_005259628.1 | 2 | 1482 | |||
GenBank mRNA | AF000561.1 | - | 2 | 590 | ||
AF097916.1 | - | 1 | 433 | |||
BC084568.1 | - | 2 | 508 | 6379572 | ||
BC113511.1 | - | 2 | 509 | 8322563 |