Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015891.2 | NP_056975.1 | 1, 2 | 206 | ||
GenBank mRNA | AB208903.1 | - | 1, 2 | 180 | ||
AF015044.1 | - | 1, 2 | 173 | |||
AF038392.1 | - | 1, 2 | 206 | |||
AF061241.1 | - | 1, 2 | 179 | |||
AK222721.1 | - | 1, 2 | 203 | |||
AK295055.1 | - | 1, 2 | 199 | |||
AK314601.1 | - | 1, 2 | 202 | |||
BC111061.1 | - | 2, 3 | 283 | |||
BC117145.1 | - | 1, 2 | 203 | 40125696 | ||
BC126114.1 | - | 1, 2 | 203 | 8991830 |