Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015976.4 | NP_057060.2 | 2 | 316 | ||
NM_152238.3 | NP_689424.2 | 2 | 316 | |||
NR_033716.1 | 3 | 436 | ||||
XM_006710677.2 | XP_006710740.1 | 2 | 333 | |||
XM_006710678.2 | XP_006710741.1 | 3 | 432 | |||
XM_011541564.2 | XP_011539866.1 | 3 | 372 | |||
XM_017001425.1 | XP_016856914.1 | 2 | 294 | |||
XM_017001426.1 | XP_016856915.1 | 3 | 431 | |||
XM_017001427.1 | XP_016856916.1 | 3 | 706 | |||
XM_017001428.1 | XP_016856917.1 | 2 | 181 | |||
GenBank mRNA | AF121857.1 | - | 2 | 159 | ||
AK025031.1 | - | 2 | 273 | |||
AK223229.1 | - | 2 | 175 | |||
AK293018.1 | - | 2 | 244 | |||
AK298211.1 | - | 2 | 162 | |||
AL049989.1 | - | 2 | 173 | |||
AL548028.3 | - | 2 | 317 | |||
BC010349.1 | - | 2 | 130 | 4093672 | ||
BC018105.1 | - | 3 | 254 | 3849824 | ||
BI856196.1 | - | 2 | 337 | |||
BT006747.1 | - | 1 | 72 |