Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244705.1 | NP_001231634.1 | 15 | 1457 | ||
NM_001244706.1 | NP_001231635.1 | 6 | 599 | |||
NM_015989.4 | NP_057073.4 | 15 | 1554 | |||
XM_011538442.2 | XP_011536744.2 | 15 | 2106 | |||
XM_011538446.2 | XP_011536748.1 | 14 | 2308 | |||
XM_011538449.1 | XP_011536751.1 | 9 | 676 | |||
XM_011538451.1 | XP_011536753.1 | 11 | 977 | |||
XM_017019416.1 | XP_016874905.1 | 12 | 1699 | |||
XM_017019417.1 | XP_016874906.1 | 12 | 1074 | |||
XM_017019418.1 | XP_016874907.1 | 11 | 1087 | |||
XM_017019419.1 | XP_016874908.1 | 10 | 931 | |||
GenBank mRNA | AB044561.1 | - | 6 | 599 | ||
AF116545.1 | - | 14 | 1297 | |||
AF116546.1 | - | 10 | 852 | |||
AF116547.1 | - | 14 | 1265 | |||
AF116548.1 | - | 15 | 1504 | |||
AK122681.1 | - | 13 | 2011 | |||
AK289659.1 | - | 10 | 973 | |||
AK315265.1 | - | 15 | 1481 | |||
BC052249.1 | - | 15 | 1457 | |||
BC098278.1 | - | 14 | 1330 | 40006057 | ||
BC098342.1 | - | 14 | 1330 | 40006051 | ||
BC099717.1 | - | 14 | 1330 | 40006058 | ||
BC105918.1 | - | 14 | 1330 | 40006053 |