Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016101.4 | NP_057185.1 | 3, 4 | 592 | ||
GenBank mRNA | AB062398.1 | - | 3, 4 | 294 | ||
AB062479.1 | - | 3, 4 | 294 | |||
AB112439.2 | - | 3, 4 | 432 | |||
AF085360.1 | - | 3, 4 | 307 | |||
AF132971.1 | - | 3, 4 | 307 | |||
AK315355.1 | - | 3, 4 | 325 | |||
BC015941.1 | - | 3, 4 | 319 | 3922688 | ||
BM754629.1 | - | 3, 4 | 460 |