Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308080.1 | NP_001295009.1 | 2 | 204 | ||
NM_016144.3 | NP_057228.1 | 2 | 177 | |||
XM_011543458.2 | XP_011541760.1 | 2 | 561 | |||
XM_011543460.2 | XP_011541762.1 | 2 | 560 | |||
XM_017009553.1 | XP_016865042.1 | 2 | 299 | |||
XM_017009555.1 | XP_016865044.1 | 2 | 559 | |||
XM_017009556.1 | XP_016865045.1 | 2 | 559 | |||
GenBank mRNA | AF078857.1 | - | 2 | 132 | ||
AF161423.1 | - | 2 | 128 | |||
AK002147.1 | - | 2 | 153 | |||
AY542165.1 | - | 2 | 100 | |||
BC005179.1 | - | 2 | 123 | 3683093 | ||
BI859871.1 | - | 2 | 193 | |||
DA001572.1 | - | 2 | 204 | |||
HY139221.1 | - | 2 | 178 |