Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318486.1 | NP_001305415.1 | 3 | 756 | ||
NM_001318489.1 | NP_001305418.1 | 4 | 698 | |||
NM_001318490.1 | NP_001305419.1 | 4 | 731 | |||
NM_001318492.1 | NP_001305421.1 | 4 | 967 | |||
NM_001318494.1 | NP_001305423.1 | 2 | 581 | |||
NM_016146.5 | NP_057230.1 | 4 | 860 | |||
GenBank mRNA | AF078862.1 | - | 4 | 790 | ||
AF105025.1 | - | 4 | 798 | |||
AF151862.1 | - | 4 | 796 | |||
AF161520.1 | - | 4 | 583 | |||
AK025623.1 | - | 4 | 563 | |||
AK290520.1 | - | 4 | 563 | |||
AK297422.1 | - | 4 | 401 | |||
BC010866.1 | - | 4 | 559 | 3876778 | ||
BF971819.1 | - | 4 | 646 | |||
BP246594.1 | - | 3 | 445 | |||
BP332308.1 | - | 4 | 434 | |||
BU624012.1 | - | 2 | 415 |