Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290007.1 | NP_001276936.1 | 10 | 1502 | ||
NM_001290008.1 | NP_001276937.1 | 9 | 1415 | |||
NM_001290009.1 | NP_001276938.1 | 9 | 2081 | |||
NM_016293.3 | NP_057377.3 | 10 | 1511 | |||
XM_005268957.3 | XP_005269014.2 | 10 | 1484 | |||
XM_005268958.3 | XP_005269015.1 | 10 | 1511 | |||
XM_017019422.1 | XP_016874911.1 | 7 | 1265 | |||
GenBank mRNA | AB032698.1 | - | 10 | 1453 | ||
AF146531.1 | - | 10 | 1423 | |||
AK000783.1 | - | 9 | 1370 | |||
AK093638.1 | - | 9 | 2008 | |||
AK225911.1 | - | 9 | 2037 | |||
AK300211.1 | - | 10 | 1490 | |||
AK314304.1 | - | 10 | 1487 | |||
BC047686.1 | - | 10 | 1477 | 5722815 |