Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUCO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282751.1 | NP_001269680.1 | 16 | 2643 | ||
NM_014283.4 | NP_055098.1 | 18 | 2803 | |||
NM_016227.3 | NP_057311.3 | 17 | 3062 | |||
XM_006711374.2 | XP_006711437.1 | 17 | 2863 | |||
XM_006711375.2 | XP_006711438.1 | 17 | 2773 | |||
XM_006711376.1 | XP_006711439.1 | 16 | 2752 | |||
GenBank mRNA | AF097535.1 | - | 17 | 3062 | ||
AJ250075.1 | - | 18 | 2801 | |||
AK302949.1 | - | 18 | 2803 | |||
AL035291.1 | - | 18 | 2704 | |||
BC137125.1 | - | 18 | 2589 | 9021122 | ||
BC144487.1 | - | 16 | 2429 |