Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other REV1 SIRNA Assays ›
Gene Symbol:
REV1
Gene Name:
REV1, DNA directed polymerase
Gene Aliases:
AIBP80, REV1L
Chromosome Location:
Chr.2: 99400475 - 99490123 on Build GRCh38
UniGene ID:
Hs.443077
Species:
Human
siRNA ID
HSS122225
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: REV1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037872.2
NP_001032961.1
9
1772
NM_001321454.1
NP_001308383.1
9
1775
NM_001321455.1
NP_001308384.1
10
1929
NM_001321458.1
NP_001308387.1
9
1772
NM_001321459.1
NP_001308388.1
10
1975
NM_001321460.1
NP_001308389.1
10
1972
NM_016316.3
NP_057400.1
9
1775
NR_135649.1
10
1972
NR_135650.1
10
1929
NR_135651.1
9
1772
NR_135652.1
9
1775
NR_135653.1
9
1775
XM_011511341.2
XP_011509643.1
2
265
XM_017004309.1
XP_016859798.1
9
2188
XM_017004310.1
XP_016859799.1
9
1554
XM_017004311.1
XP_016859800.1
9
1554
XM_017004312.1
XP_016859801.1
9
1767
XM_017004313.1
XP_016859802.1
9
3501
XM_017004314.1
XP_016859803.1
5
1025
XM_017004315.1
XP_016859804.1
5
1028
GenBank mRNA
AB047646.1
-
9
1697
AF151538.1
-
9
1728
AF206019.1
-
9
1737
AF357886.1
-
9
1693
AJ131720.1
-
1
94
AK025176.1
-
1
54
AK307719.1
-
9
1686
BC037734.1
-
10
1910
4824689
BC130411.1
-
8
1543
40146442
BP332286.1
-
3
239
DA304601.1
-
4
436
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
DNA replication
response to UV
translesion synthesis
error-prone translesion synthesis
damaged DNA binding
protein binding
deoxycytidyl transferase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline