Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256539.3 | NP_001243468.2 | 1 | 448 | ||
NM_001256540.3 | NP_001243469.2 | 1 | 448 | |||
NM_001291305.2 | NP_001278234.1 | 1 | 448 | |||
NM_001291307.2 | NP_001278236.1 | 1 | 448 | |||
NM_001291308.2 | NP_001278237.1 | 1 | 448 | |||
NM_001317975.1 | NP_001304904.1 | 1 | 448 | |||
NM_016391.7 | NP_057475.2 | 1 | 448 | |||
NR_134316.1 | 1 | 448 | ||||
NR_134318.1 | 1 | 448 | ||||
XM_011534567.1 | XP_011532869.1 | 1 | 127 | |||
GenBank mRNA | AF151019.1 | - | 1 | 446 | ||
AF151875.1 | - | 2 | 516 | |||
AF161460.1 | - | 1 | 122 | |||
AK300883.1 | - | 1 | 127 | |||
AK303281.1 | - | 1 | 119 | |||
AW837047.1 | - | 1 | 97 | |||
AY561704.1 | - | 1 | 60 | |||
BC019331.1 | - | 1 | 283 | 3534019 | ||
BC032424.2 | - | 1 | 91 | 5416724 | ||
BC040106.1 | - | 1 | 108 | 6066208 | ||
BP234677.1 | - | 1 | 119 | |||
BU594763.1 | - | 1 | 122 | |||
CB122385.1 | - | 1 | 108 | |||
CR457169.1 | - | 1 | 60 | |||
HY064071.1 | - | 1 | 131 |