Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317975.1 | NP_001304904.1 | 4 | 931 | ||
NM_016391.7 | NP_057475.2 | 5 | 842 | |||
NR_134316.1 | 4 | 948 | ||||
XM_011534567.1 | XP_011532869.1 | 4 | 451 | |||
GenBank mRNA | AF151019.1 | - | 5 | 837 | ||
AF151875.1 | - | 6 | 910 | |||
AF161460.1 | - | 5 | 516 | |||
AY561704.1 | - | 5 | 454 | |||
BC040106.1 | - | 5 | 502 | 6066208 | ||
BM988284.1 | - | 1 | 391 | |||
CR457169.1 | - | 5 | 454 |