Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256539.3 | NP_001243468.2 | 4, 5 | 791 | ||
NM_001317975.1 | NP_001304904.1 | 4 | 968 | |||
NM_016391.7 | NP_057475.2 | 5 | 879 | |||
NR_134316.1 | 4 | 985 | ||||
XM_011534567.1 | XP_011532869.1 | 4 | 488 | |||
GenBank mRNA | AF151019.1 | - | 5 | 874 | ||
AF151875.1 | - | 6 | 947 | |||
AF161460.1 | - | 5 | 553 | |||
AY561704.1 | - | 5 | 491 | |||
BC032424.2 | - | 4, 5 | 434 | 5416724 | ||
BC040106.1 | - | 5 | 539 | 6066208 | ||
BM988284.1 | - | 1 | 354 | |||
CB122385.1 | - | 4, 5 | 451 | |||
CR457169.1 | - | 5 | 491 |