Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198976.2 | NP_945327.2 | 13 | 1585 | ||
GenBank mRNA | AF161479.1 | - | 13 | 1515 | ||
AJ238374.1 | - | 8 | 982 | |||
AJ238375.1 | - | 8 | 908 | |||
AJ238379.1 | - | 13 | 1519 | |||
AK001316.1 | - | 13 | 1493 | |||
AK023927.1 | - | 13 | 1510 | |||
AK127431.1 | - | 3 | 2517 | |||
AK293410.1 | - | 13 | 1537 | |||
BC014952.1 | - | 13 | 1521 | 4860894 | ||
BX647417.1 | - | 13 | 1520 |