Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTSE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_016426.6 | NP_057510.4 | 11 | 2254 | ||
XM_005261627.3 | XP_005261684.1 | 11 | 3114 | |||
XM_011530211.2 | XP_011528513.1 | 10 | 3056 | |||
XM_017028815.1 | XP_016884304.1 | 10 | 2504 | |||
XM_017028816.1 | XP_016884305.1 | 10 | 2914 | |||
GenBank mRNA | AF223408.1 | - | 10 | 2055 | ||
AK025563.1 | - | 11 | 2081 | |||
AK222806.1 | - | 11 | 2081 | |||
AK303087.1 | - | 10 | 2025 | |||
AK315550.1 | - | 11 | 2080 | |||
AY927425.1 | - | LIT3464 AY927425 |
1 | 368 | ||
BC006325.2 | - | 10 | 2032 | 4138532 | ||
BC075828.1 | - | 11 | 2155 | 5092448 | ||
CT841519.1 | - | 10 | 1998 |