Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3HC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282190.1 | NP_001269119.1 | 7 | 867 | ||
NM_001282191.1 | NP_001269120.1 | 6 | 821 | |||
NM_016478.4 | NP_057562.3 | 6 | 821 | |||
XM_005250403.2 | XP_005250460.1 | 4 | 481 | |||
XM_011516288.2 | XP_011514590.1 | 5 | 647 | |||
XM_011516289.2 | XP_011514591.1 | 5 | 1032 | |||
XM_011516290.2 | XP_011514592.1 | 4 | 2371 | |||
XM_017012287.1 | XP_016867776.1 | 6 | 816 | |||
XM_017012288.1 | XP_016867777.1 | 3 | 1323 | |||
GenBank mRNA | AF151050.1 | - | 6 | 742 | ||
AJ537494.1 | - | 6 | 736 | |||
AK001317.1 | - | 7 | 790 | |||
AK022373.1 | - | 6 | 744 | |||
BC000190.1 | - | 6 | 729 | |||
BC011551.1 | - | 6 | 737 | 4561164 | ||
BC028917.1 | - | 1 | 13 |