Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3HC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282190.1 | NP_001269119.1 | 8 | 993 | ||
NM_001282191.1 | NP_001269120.1 | 7 | 947 | |||
NM_016478.4 | NP_057562.3 | 7 | 947 | |||
XM_005250403.2 | XP_005250460.1 | 5 | 607 | |||
XM_011516288.2 | XP_011514590.1 | 6 | 773 | |||
XM_011516289.2 | XP_011514591.1 | 6 | 1158 | |||
XM_011516290.2 | XP_011514592.1 | 5 | 2497 | |||
XM_017012288.1 | XP_016867777.1 | 4 | 1449 | |||
GenBank mRNA | AF151050.1 | - | 7 | 868 | ||
AJ537494.1 | - | 7 | 862 | |||
AK001317.1 | - | 8 | 916 | |||
AK022373.1 | - | 7 | 870 | |||
BC000190.1 | - | 7 | 855 | |||
BC011551.1 | - | 7 | 863 | 4561164 | ||
BC028917.1 | - | 2 | 139 |