Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPHLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143787.1 | NP_001137259.1 | 5 | 483 | ||
NM_016488.6 | NP_057572.5 | 6 | 648 | |||
NM_201438.1 | NP_958846.1 | 6 | 526 | |||
NM_201439.1 | NP_958847.1 | 6 | 648 | |||
NM_201440.1 | NP_958848.1 | 6 | 526 | |||
NM_201515.1 | NP_958923.1 | 7 | 691 | |||
XM_011538459.2 | XP_011536761.1 | 7 | 669 | |||
XM_011538467.2 | XP_011536769.1 | 6 | 508 | |||
XM_017019434.1 | XP_016874923.1 | 7 | 669 | |||
XM_017019435.1 | XP_016874924.1 | 7 | 668 | |||
XM_017019436.1 | XP_016874925.1 | 7 | 669 | |||
XM_017019437.1 | XP_016874926.1 | 6 | 626 | |||
XM_017019438.1 | XP_016874927.1 | 6 | 626 | |||
XM_017019439.1 | XP_016874928.1 | 7 | 669 | |||
XM_017019444.1 | XP_016874933.1 | 6 | 508 | |||
XM_017019445.1 | XP_016874934.1 | 5 | 502 | |||
XM_017019446.1 | XP_016874935.1 | 5 | 501 | |||
GenBank mRNA | AF151040.1 | - | 6 | 489 | ||
AK055690.1 | - | 6 | 636 | |||
AK129910.1 | - | 7 | 691 | |||
AK303612.1 | - | 5 | 483 | |||
AY039238.1 | - | 6 | 630 | |||
BC025306.1 | - | 5 | 450 | |||
BC039832.1 | - | 6 | 493 | 6051282 | ||
BC040449.1 | - | 6 | 493 | |||
CF242917.1 | - | 5 | 498 |