Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324066.1 | NP_001310995.1 | 5 | 901 | ||
NM_001324067.1 | NP_001310996.1 | 4 | 812 | |||
NM_016641.3 | NP_057725.1 | 5 | 888 | |||
NR_136689.1 | 5 | 937 | ||||
GenBank mRNA | AF212862.1 | - | 5 | 844 | ||
AF306698.1 | - | 5 | 797 | |||
AK312912.1 | - | 5 | 858 | |||
AY463154.1 | - | 5 | 724 | |||
BC014981.1 | - | 5 | 779 | 4855079 | ||
BC025273.1 | - | 5 | 839 | 5427491 |