Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324066.1 | NP_001310995.1 | 5 | 957 | ||
NM_001324067.1 | NP_001310996.1 | 4 | 868 | |||
NM_016641.3 | NP_057725.1 | 5 | 944 | |||
NR_136689.1 | 5 | 993 | ||||
GenBank mRNA | AF212862.1 | - | 5 | 900 | ||
AF306698.1 | - | 5 | 853 | |||
AK312912.1 | - | 5 | 914 | |||
AY463154.1 | - | 5 | 780 | |||
BC014981.1 | - | 5 | 835 | 4855079 | ||
BC025273.1 | - | 5 | 895 | 5427491 |