Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LARP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267039.1 | NP_001253968.1 | 13 | 1963 | ||
NM_015454.2 | NP_056269.1 | 11 | 1627 | |||
NM_016648.3 | NP_057732.2 | 11 | 1755 | |||
NR_049768.1 | 11 | 1652 | ||||
GenBank mRNA | AF068284.1 | - | 10 | 904 | ||
AK000274.1 | - | 11 | 1565 | |||
AK225900.1 | - | 8 | 1124 | |||
AL049996.1 | - | 5 | 602 | |||
BC066945.1 | - | 13 | 1801 | 4837780 | ||
BC107709.1 | - | 5 | 596 | |||
BX647100.1 | - | 11 | 1489 | |||
CA314598.1 | - | 3 | 545 | |||
EU667388.1 | - | 11 | 1498 |