Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AZIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301668.1 | NP_001288597.1 | 6 | 1155 | ||
NM_015878.5 | NP_056962.2 | 7 | 1301 | |||
NM_148174.3 | NP_680479.1 | 6 | 1155 | |||
XM_005250969.1 | XP_005251026.1 | 6 | 1116 | |||
XM_011517126.1 | XP_011515428.1 | 7 | 1288 | |||
XM_011517127.1 | XP_011515429.1 | 7 | 1279 | |||
XM_011517128.2 | XP_011515430.1 | 6 | 1299 | |||
XM_017013563.1 | XP_016869052.1 | 7 | 1267 | |||
XM_017013564.1 | XP_016869053.1 | 7 | 1279 | |||
XM_017013565.1 | XP_016869054.1 | 7 | 1418 | |||
XM_017013566.1 | XP_016869055.1 | 6 | 1299 | |||
GenBank mRNA | AK095493.1 | - | 6 | 1083 | ||
AK290516.1 | - | 7 | 1033 | |||
BC013420.2 | - | 7 | 1240 | 3509827 | ||
BC019279.1 | - | 6 | 1102 | 2823000 | ||
BC042404.1 | - | 6 | 1126 | |||
BT006858.1 | - | 4 | 528 | |||
CR456745.1 | - | 4 | 528 | |||
D88674.1 | - | 7 | 1237 |