Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014433.2 | NP_001014433.1 | 6 | 593 | ||
NM_001014837.1 | NP_001014837.1 | 6 | 842 | |||
NM_001014838.1 | NP_001014838.1 | 6 | 811 | |||
NM_001014840.1 | NP_001014840.1 | 6 | 721 | |||
NM_015921.2 | NP_057005.1 | 5 | 984 | |||
GenBank mRNA | AF106943.1 | - | 5 | 621 | ||
AF230924.1 | - | 5 | 407 | |||
AK311556.1 | - | 1 | 1351 | |||
BC005890.2 | - | 6 | 418 | 3505574 | ||
BC107751.1 | - | 5 | 621 | 6714433 |