Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014433.2 | NP_001014433.1 | 6 | 670 | ||
NM_001014837.1 | NP_001014837.1 | 6 | 919 | |||
NM_001014838.1 | NP_001014838.1 | 6 | 888 | |||
NM_001014840.1 | NP_001014840.1 | 6 | 798 | |||
NM_015921.2 | NP_057005.1 | 5 | 1061 | |||
GenBank mRNA | AF106943.1 | - | 5 | 698 | ||
AF230924.1 | - | 5 | 484 | |||
BC005890.2 | - | 6 | 495 | 3505574 | ||
BC107751.1 | - | 5 | 698 | 6714433 |