Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015934.4 | NP_057018.1 | 9 | 1108 | ||
GenBank mRNA | AF123534.1 | - | 9 | 882 | ||
AF161469.1 | - | 7 | 761 | |||
AF263608.1 | - | 9 | 882 | |||
AK023975.1 | - | 8 | 807 | |||
AK307396.1 | - | 9 | 1088 | |||
AL117554.1 | - | 9 | 1091 | |||
BC001707.1 | - | 9 | 1074 | |||
BC009306.2 | - | 9 | 1038 | |||
BC018805.1 | - | 9 | 1061 | |||
BC032592.2 | - | 9 | 1032 | 5527148 |