Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007239.1 | NP_001007240.1 | 4 | 1066 | ||
NM_014955.2 | NP_055770.1 | 4 | 1209 | |||
NM_015935.4 | NP_057019.3 | 4 | 1534 | |||
XM_017001455.1 | XP_016856944.1 | 3 | 1028 | |||
XM_017001456.1 | XP_016856945.1 | 3 | 1028 | |||
XM_017001457.1 | XP_016856946.1 | 3 | 1028 | |||
XM_017001458.1 | XP_016856947.1 | 4 | 1651 | |||
XM_017001459.1 | XP_016856948.1 | 4 | 1201 | |||
GenBank mRNA | AB020666.2 | - | 4 | 1289 | ||
AB242174.1 | - | 4 | 1459 | |||
AB242175.1 | - | 4 | 1447 | |||
AF132936.1 | - | 4 | 1459 | |||
AK001172.1 | - | 2 | 221 | |||
AK027552.1 | - | 4 | 1453 | |||
AK027621.1 | - | 4 | 1148 | |||
AK027886.1 | - | 3 | 1005 | |||
AK129561.1 | - | 3 | 882 | |||
AK223493.1 | - | 4 | 1495 | |||
AK291740.1 | - | 4 | 1481 | |||
AK304471.1 | - | 5 | 1315 | |||
AL049669.1 | - | 4 | 991 | |||
BC006814.1 | - | 1 | 84 | |||
BC029083.1 | - | 4 | 1288 | 4509086 |