Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUC7L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244585.1 | NP_001231514.1 | 6, 7 | 690 | ||
NM_001270643.1 | NP_001257572.1 | 6, 7 | 1002 | |||
NM_016019.4 | NP_057103.2 | 5, 6 | 931 | |||
GenBank mRNA | AF151817.1 | - | 5, 6 | 490 | ||
AF151832.1 | - | 5, 6 | 515 | |||
AK001476.1 | - | 5, 6 | 823 | |||
AK022895.1 | - | 5, 6 | 806 | |||
AK092044.1 | - | 4, 5 | 1312 | |||
AK093864.1 | - | 6, 7 | 789 | |||
AK095839.1 | - | 4, 5 | 1843 | |||
AK298166.1 | - | 6, 7 | 690 | |||
BC017163.2 | - | 5, 6 | 848 | 3920377 | ||
BC042625.1 | - | 6, 7 | 936 | 5178074 | ||
BC050708.2 | - | 5, 6 | 861 | 6500791 | ||
BC056886.1 | - | 5, 6 | 806 | 6147116 |
Gene Symbol: C7orf55-LUC7L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244584.2 | NP_001231513.1 | 6, 7 | 966 | ||
GenBank mRNA | AK297642.1 | - | 6, 7 | 738 |