Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322280.1 | NP_001309209.1 | 3 | 390 | ||
NM_001322281.1 | NP_001309210.1 | 3 | 539 | |||
NM_001322282.1 | NP_001309211.1 | 3 | 539 | |||
NM_016029.3 | NP_057113.1 | 3 | 539 | |||
XM_017021368.1 | XP_016876857.1 | 3 | 647 | |||
GenBank mRNA | AF126782.1 | - | 485 | |||
AF151844.1 | - | 3 | 432 | |||
AK313285.1 | - | 3 | 464 | |||
AL117420.1 | - | 3 | 475 | |||
AY359031.1 | - | 3 | 348 | |||
BC000637.2 | - | 3 | 375 | 3344010 | ||
BC007337.2 | - | 3 | 393 | 3616802 |