Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318498.1 | NP_001305427.1 | 6 | 471 | ||
NM_001318499.1 | NP_001305428.1 | 6 | 536 | |||
NM_016055.5 | NP_057139.1 | 5 | 413 | |||
NR_134658.1 | 4 | 375 | ||||
NR_134659.1 | 7 | 666 | ||||
NR_134660.1 | 5 | 531 | ||||
NR_134661.1 | 4 | 324 | ||||
XM_017017919.1 | XP_016873408.1 | 6 | 506 | |||
GenBank mRNA | AF151876.1 | - | 5 | 969 | ||
AF161408.1 | - | 5 | 416 | |||
AK297746.1 | - | 6 | 471 | |||
AK309219.1 | - | 4 | 353 | |||
AK310005.1 | - | 7 | 645 | |||
BC009764.1 | - | 5 | 406 | 4093230 | ||
BI549412.1 | - | 4 | 367 | |||
BM817807.1 | - | 5 | 411 | |||
BM828996.1 | - | 5 | 412 | |||
BP210564.1 | - | 6 | 514 | |||
BQ446725.1 | - | 4 | 581 | |||
BU932890.1 | - | 6 | 537 | |||
BX354271.2 | - | 5 | 496 | |||
CF541319.1 | - | 3 | 472 | |||
DA549653.1 | - | 6 | 471 |