Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK5RAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278167.1 | NP_001265096.1 | 8 | 1129 | ||
NM_001278169.1 | NP_001265098.1 | 8 | 897 | |||
NM_016082.4 | NP_057166.4 | 8 | 981 | |||
NM_016408.3 | NP_057492.2 | 8 | 1129 | |||
XM_011528856.2 | XP_011527158.1 | 5 | 1865 | |||
XM_011528857.2 | XP_011527159.1 | 7 | 741 | |||
XM_011528858.1 | XP_011527160.1 | 7 | 980 | |||
XM_011528859.2 | XP_011527161.1 | 7 | 740 | |||
XM_011528860.1 | XP_011527162.1 | 5 | 440 | |||
XM_017027876.1 | XP_016883365.1 | 7 | 879 | |||
XM_017027877.1 | XP_016883366.1 | 2 | 193 | |||
GenBank mRNA | AF152097.1 | - | 9 | 1107 | ||
AF161516.1 | - | 8 | 1077 | |||
AK023992.1 | - | 8 | 1079 | |||
AK027449.1 | - | 8 | 929 | |||
AK222745.1 | - | 8 | 1079 | |||
AK292075.1 | - | 8 | 847 | |||
AK295342.1 | - | 8 | 1065 | |||
AK314532.1 | - | 8 | 1079 | |||
AY462283.1 | - | 8 | 1080 | |||
AY462284.1 | - | 8 | 1080 | |||
BC050706.1 | - | 8 | 1072 | 4418659 | ||
CX785345.1 | - | 3 | 336 | |||
DR156307.1 | - | 7 | 738 | |||
KU178645.1 | - | 7 | 967 |