Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016084.4 | NP_057168.1 | 2 | 520 | ||
GenBank mRNA | AF069506.1 | - | 2 | 451 | ||
AF172846.1 | - | 2 | 369 | |||
AF177335.1 | - | 2 | 499 | |||
AF498923.1 | - | 2 | 306 | |||
AK312796.1 | - | 2 | 520 | |||
BC018041.1 | - | 2 | 518 | 4797482 | ||
BC042688.1 | - | 2 | 486 |