Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STYXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317785.1 | NP_001304714.1 | 8 | 1067 | ||
NM_001317786.1 | NP_001304715.1 | 8 | 917 | |||
NM_001317787.1 | NP_001304716.1 | 6 | 626 | |||
NM_001317789.1 | NP_001304718.1 | 6 | 576 | |||
NM_016086.2 | NP_057170.1 | 8 | 1064 | |||
XM_011516293.2 | XP_011514595.1 | 8 | 910 | |||
XM_011516300.1 | XP_011514602.1 | 7 | 720 | |||
XM_017012296.1 | XP_016867785.1 | 9 | 1024 | |||
XM_017012300.1 | XP_016867789.1 | 9 | 944 | |||
XM_017012301.1 | XP_016867790.1 | 7 | 764 | |||
XM_017012302.1 | XP_016867791.1 | 7 | 767 | |||
GenBank mRNA | AA782050.1 | - | 2 | 353 | ||
AF069762.1 | - | 8 | 1060 | |||
AF188202.1 | - | 4 | 298 | |||
AF188204.1 | - | 5 | 441 | |||
AK308912.1 | - | 8 | 891 | |||
AL533027.3 | - | 8 | 865 | |||
BC024035.2 | - | 8 | 1027 | 2988900 | ||
BM972220.1 | - | 2 | 366 | |||
BX537896.1 | - | 6 | 606 |